Sodium-3-(4-hydroxy-3-methoxyphenyl)lactate ≥99.0% (by HPLC)

Supplier: TCI America

H0538-1G H0538-100MG
TCH0538-1GEA 1139.18 CAD
TCH0538-1G TCH0538-100MG
Sodium-3-(4-hydroxy-3-methoxyphenyl)lactate ≥99.0% (by HPLC)
Sodium-3-(4-hydroxy-3-methoxyphenyl)lactate

CAS Number: 77305-02-3
MDL Number: MFCD00056719
Molecular Formula: C10H12O5
Molecular Weight: 234.18
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 3-(4-Hydroxy-3-methoxyphenyl)lactic Acid Sodium Salt, Sodium 2-Hydroxy-3-(4-hydroxy-3-methoxyphenyl)propionate, 2-Hydroxy-3-(4-hydroxy-3-methoxyphenyl)propionic Acid Sodium Salt

Formula: C₁₀H₁₂O₅
MDL Number: MFCD00056719
CAS Number: 77305-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=baae76430cf32b66fe33b798e670d700a812e232&vid=baae76430cf32b66fe33b798e670d700a812e232&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR