1-Heptyloxy-4-nitrobenzene

Supplier: TCI America

H0435-5ML
TCH0435-005MLEA 340.49 CAD
TCH0435-005ML
1-Heptyloxy-4-nitrobenzene
1-Heptyloxy-4-nitrobenzene

CAS Number: 13565-36-1
MDL Number: MFCD00024692
Molecular Formula: C13H19NO3
Molecular Weight: 237.30
Form: Clear Liquid
Color: Yellow
Boiling point (°C): 203
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Heptyl 4-Nitrophenyl Ether

Formula: C₁₃H₁₉NO₃
Boiling Pt: 203 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00024692
CAS Number: 13565-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR