2-Quinolinol ≥98.0%

Supplier: TCI America
Warning

H0304-1G H0304-10G
TCH0304-001GEA 31.6 CAD
TCH0304-001G TCH0304-010G
2-Quinolinol ≥98.0%
2-Quinolinol

CAS Number: 59-31-4
MDL Number: MFCD00006743
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 197


Other Product Names / Synonyms: Carbostyril, 2-Hydroxyquinoline

Formula: C₉H₇NO
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 197 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006743
CAS Number: 59-31-4
EINECS: 200-420-6
Merck Index: 13,01835
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=680efe8f2dbf03e8dd0075c70ab71b589f1e668c&vid=bbfc697e64510df892015b19092822fbff00a86a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR