Sodium-DL-3-hydroxybutyrate ≥99.0%

Supplier: TCI America

Synonyms: Sodium 3-hydroxybutanoate, DL-3-Hydroxybutyric acid Sodium salt

H0231-25G
TCH0231-025GEA 144.14 CAD
TCH0231-025G
Sodium-DL-3-hydroxybutyrate ≥99.0%
Sodium-DL-3-hydroxybutyrate

CAS Number: 150-83-4
MDL Number: MFCD00016716
Molecular Formula: C4H8O3
Molecular Weight: 126.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 170


Other Product Names / Synonyms: DL-3-Hydroxybutyric Acid Sodium Salt

Formula: C₄H₈O₃
MW: 126.09 g/mol
Melting Pt: 170 °C
MDL Number: MFCD00016716
CAS Number: 150-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=117df6186d33918fbdc7c7b6b6a6378d063553e7&vid=117df6186d33918fbdc7c7b6b6a6378d063553e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR