Ethyl-3-hydroxybutyrate ≥98.0%

Supplier: TCI America

H0230-500G H0230-25G
TCH0230-500GEA 312.84 CAD
TCH0230-500G TCH0230-025G
Ethyl-3-hydroxybutyrate ≥98.0%
Ethyl 3-hydroxybutyrate

CAS Number: 5405-41-4
MDL Number: MFCD00004545
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 64
Specific Gravity (20/20): 1.02


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: DL-3-Hydroxybutyric Acid Ethyl Ester

Formula: C₆H₁₂O₃
Boiling Pt: 180 °C
Density: 1.02 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00004545
CAS Number: 5405-41-4
EINECS: 226-456-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70361ea9578ed11b244549ec9980c2c59d68565f&vid=70361ea9578ed11b244549ec9980c2c59d68565f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR