1-Heptanol ≥98.0%

Supplier: TCI America
Warning

H0033-500ML H0033-25ML
TCH0033-500MLEA 64.78 CAD
TCH0033-500ML TCH0033-25ML
1-Heptanol ≥98.0%
1-Heptanol

CAS Number: 111-70-6
MDL Number: MFCD00002986
Molecular Formula: C7H16O
Molecular Weight: 116.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Melting point (°C): -34
Flash Point (°C): 71
Specific Gravity (20/20): 0.82


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Heptyl Alcohol

Formula: H₃C(CH₂)₅CH₂OH
Boiling Pt: 177 °C
Melting Pt: –34 °C
Density: 0.82 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9
Merck Index: 13,04681
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d94fb43809b000bdd5da7153a0758c73437d766&vid=17a1cce2592226e10b4f96f56b5eabbd912d2745&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR