D-(-)-Glutamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

G0057-250G G0057-25G
TCG0057-250GEA 191.18 CAD
TCG0057-250G TCG0057-025G
D-(-)-Glutamic acid ≥98.0% (by titrimetric analysis)
D(-)-Glutamic acid

CAS Number: 6893-26-1
MDL Number: MFCD00063112
Molecular Formula: C5H9NO4
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Glu-OH

Formula: C₅H₉NO₄
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 213 °C
Density: 1.533 g/cm³ (20 °C)
MDL Number: MFCD00063112
CAS Number: 6893-26-1
EINECS: 230-000-8
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb48bb7a6c8637bd30e8cd976bc918253026dbd4&vid=fb48bb7a6c8637bd30e8cd976bc918253026dbd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR