4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic acid ≥97.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 4-Fmoc-morpholine-3-carboxylic acid

F1050-200MG
TCF1050-200MGEA 325.48 CAD
TCF1050-200MG
4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic acid ≥97.0% (by HPLC)
4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic acid
Formula: C₂₀H₁₉NO₅
MW: 353.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01632020
CAS Number: 204320-51-4

Order Now

Specification Test Results

Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR