5-Fluoro-2-methylindole ≥98.0% (by GC)

Supplier: TCI America
Warning

F1047-1G F1047-5G
TCF1047-1GEA 219.62 CAD
TCF1047-1G TCF1047-5G
5-Fluoro-2-methylindole ≥98.0% (by GC)
5-Fluoro-2-methylindole
Formula: C9H8FN
MW: 149.17 g/mol
Boiling Pt: 120°C
MDL Number: MFCD02093649
CAS Number: 399-72-4

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR