3-Formyl-6-methylchromone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6-Methyl-4-oxo-4H-chromene-3-carbaldehyde

F1005-1G F1005-5G
TCF1005-1GEA 36.34 CAD
TCF1005-1G TCF1005-5G
3-Formyl-6-methylchromone ≥98.0% (by GC)
3-Formyl-6-methylchromone
Formula: C₁₁H₈O₃
MDL Number: MFCD00138943
CAS Number: 42059-81-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR