1-Fluoro-3,3-dimethyl-1,2-benziodoxole ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

F0957-1G
TCF0957-1GEA 328.96 CAD
TCF0957-1G
1-Fluoro-3,3-dimethyl-1,2-benziodoxole ≥98.0% (by titrimetric analysis)
1-Fluoro-3,3-dimethyl-1,2-benziodoxole
Formula: C₉H₁₀FIO
MDL Number: MFCD28397076
CAS Number: 1391728-13-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystaline
Purity (Iodometric Titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR