3-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (3-Fluoro-2-methylphenyl)boronic acid, 3-Fluoro-2-methylbenzeneboronic acid

F0955-1G
TCF0955-1GEA 170.32 CAD
TCF0955-1G
3-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-2-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
MDL Number: MFCD07368679
CAS Number: 163517-61-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR