Fluorescein-5-maleimide ≥97.0% (by HPLC)

Supplier: TCI America

F0810-25MG
TCF0810-25MGEA 476.7 CAD
TCF0810-25MG
Fluorescein-5-maleimide ≥97.0% (by HPLC)
Fluorescein-5-maleimide

CAS Number: 75350-46-8
MDL Number: MFCD00077345
Molecular Formula: C24H13NO7
Molecular Weight: 427.37
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Fluorometry & Fluorescent Imaging, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 5-Maleimidofluorescein

Formula: C₂₄H₁₃NO₇
MDL Number: MFCD00077345
CAS Number: 75350-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=198fa6f64954047a177228ddc58201de7de94c03&vid=198fa6f64954047a177228ddc58201de7de94c03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR