Fluorescein 6-isothiocyanate (isomer II) ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3',6'-Dihydroxy-6-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 6-FITC, Fluorescein Isothiocyanate, 6-Isothiocyanatofluorescein, Fluorescein-6-isothiocyanate

F0783-100MG
TCF0783-100MGEA 462.78 CAD
TCF0783-100MG
Fluorescein 6-isothiocyanate (isomer II) ≥97.0% (by HPLC, titration analysis)
Fluorescein 6-isothiocyanate (isomer II)

CAS Number: 18861-78-4
MDL Number: MFCD00041838
Molecular Formula: C21H11NO5S
Molecular Weight: 389.38
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Stains & Dyes, Fluorometry & Fluorescent Imaging, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 6-Isothiocyanatofluorescein, Fluorescein Isothiocyanate, 6-FITC

Formula: C₂₁H₁₁NO₅S
MW: 389.38 g/mol
MDL Number: MFCD00041838
CAS Number: 18861-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64579af4d55460d289ea475ce2afc21582263db9&vid=64579af4d55460d289ea475ce2afc21582263db9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR