3-Fluoro-4-hydroxybenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

F0725-1G F0725-5G
TCF0725-1GEA 134.3 CAD
TCF0725-1G TCF0725-5G
3-Fluoro-4-hydroxybenzaldehyde ≥98.0% (by GC)
3-Fluoro-4-hydroxybenzaldehyde

CAS Number: 405-05-0
MDL Number: MFCD00016626
Molecular Formula: C7H5FO2
Molecular Weight: 140.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125

Formula: C₇H₅FO₂
Melting Pt: 125 °C
MDL Number: MFCD00016626
CAS Number: 405-05-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7aa4647e7860d1722eaa8ebabb4d511d68f6ebd4&vid=7aa4647e7860d1722eaa8ebabb4d511d68f6ebd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR