1,4-Dimethoxy-2-fluorobenzene ≥97.0%

Supplier: TCI America

Synonyms: 2-Fluoro-1,4-dimethoxybenzene

F0633-5G
TCF0633-5GEA 153.73 CAD
TCF0633-5G
1,4-Dimethoxy-2-fluorobenzene ≥97.0%
1,4-Dimethoxy-2-fluorobenzene

CAS Number: 82830-49-7
MDL Number: MFCD00042487
Molecular Formula: C8H9FO2
Molecular Weight: 156.16
Purity/Analysis Method: >97.0% (GC)
Boiling point (°C): 121
Freezing point (°C): 24

Formula: C₈H₉FO₂
MW: 156.16 g/mol
Boiling Pt: 121 °C
MDL Number: MFCD00042487
CAS Number: 82830-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b5c72a5cd8c7a151bf1467f4bafa97d0ee5ee09&vid=3b5c72a5cd8c7a151bf1467f4bafa97d0ee5ee09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR