2-(4-Fluorophenyl)ethanol ≥97.0%

Supplier: TCI America

F0624-5G
TCF0624-5GEA 93.22 CAD
TCF0624-5G
2-(4-Fluorophenyl)ethanol ≥97.0%
2-(4-Fluorophenyl)ethanol

CAS Number: 7589-27-7
MDL Number: MFCD00002898
Molecular Formula: C8H9FO
Molecular Weight: 140.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 86
Specific Gravity (20/20): 1.13

Formula: C₈H₉FO
Boiling Pt: 118 °C
Density: 1.13 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00002898
CAS Number: 7589-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d494efd61ccfd55b1126e939cf42ecd071c4892e&vid=d494efd61ccfd55b1126e939cf42ecd071c4892e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR