Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Fmoc-Ser-OtBu, (S)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate, Nɑ-Fmoc-L-serine tert-Butyl Ester

F0516-5G F0516-1G
TCF0516-5GEA 1434.64 CAD
TCF0516-5G TCF0516-1G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester

CAS Number: 110797-35-8
Molecular Formula: C22H25NO5
Molecular Weight: 383.44
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 127
Specific rotation [a]20/D: 7 deg (C=1, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-L-serine tert-Butyl Ester, Fmoc-Ser-OtBu

Formula: C₂₂H₂₅NO₅
MW: 383.44 g/mol
Melting Pt: 127 °C
MDL Number: MFCD01861331
CAS Number: 110797-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e7e5dd87100c36d6d63edcf1d3e3855b083039f&vid=5e7e5dd87100c36d6d63edcf1d3e3855b083039f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR