2-Fluoro-4-iodoaniline ≥98.0%

Supplier: TCI America

F0417-25G F0417-5G
TCF0417-025GEA 109.02 CAD
TCF0417-025G TCF0417-005G
2-Fluoro-4-iodoaniline ≥98.0%
2-Fluoro-4-iodoaniline

CAS Number: 29632-74-4
MDL Number: MFCD00011738
Molecular Formula: C6H5FIN
Molecular Weight: 237.02
Purity/Analysis Method: >98.0% (GC)
Melting point (°C): 55
Flash Point (°C): 113

Formula: C₆H₅FIN
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 55 °C
Flash Pt: 113 °C
MDL Number: MFCD00011738
CAS Number: 29632-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18c6d4cb70029784381d1997881b8a21aff4f23d&vid=18c6d4cb70029784381d1997881b8a21aff4f23d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR