2-[2-(Furan-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

F0362-5G
TCF0362-005GEA 158 CAD
TCF0362-005G
2-[2-(Furan-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine ≥95.0% (by HPLC, total nitrogen)
2-[2-(Furan-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine

CAS Number: 154880-05-4
MDL Number: MFCD00671535
Molecular Formula: C11H5Cl6N3O
Molecular Weight: 407.88
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal
Melting point (°C): 142


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 2,4-Bis(trichloromethyl)-6-[2-(furan-2-yl)vinyl]-1,3,5-triazine

Formula: C₁₁H₅Cl₆N₃O
Melting Pt: 142 °C
MDL Number: MFCD00671535
CAS Number: 154880-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859a58253192f18a1d24d6ef4032095f2fbb5ebd&vid=859a58253192f18a1d24d6ef4032095f2fbb5ebd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR