N-Formyl-L-alanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: L-Alanine, N-formyl-, (2S)-2-formamidopropanoic acid, For-Ala-OH

F0108-1G
TCF0108-001GEA 214.88 CAD
TCF0108-001G
N-Formyl-L-alanine ≥98.0% (by titrimetric analysis)
N-Formyl-L-alanine

CAS Number: 10512-86-4
MDL Number: MFCD00037357
Molecular Formula: C4H7NO3
Molecular Weight: 117.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -32 deg (C=4, EtOH)

Formula: C₄H₇NO₃
MW: 117.10 g/mol
MDL Number: MFCD00037357
CAS Number: 10512-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a2f1d7ba81fa80d070591693a98d40f407e115d&vid=6a2f1d7ba81fa80d070591693a98d40f407e115d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR