Ethyl (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a']dinaphthalene-4-carboxylate ≥97.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a']dinaphthalene-4-carboxylic acid Ethyl ester

E1267-50MG
TCE1267-50MGEA 500.86 CAD
TCE1267-50MG
Ethyl (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a']dinaphthalene-4-carboxylate ≥97.0% (by HPLC)
Ethyl (11bR)-4-Amino-2,6-bis(3,5-di-tert-butylphenyl)-4,5-dihydro-3H-cyclohepta[1,2-a:7,6-a']dinaphthalene-4-carboxylate
Formula: C₅₄H₆₃NO₂
Storage Temperature: Refrigerator
CAS Number: 1678540-23-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR