Ethyl-6-aminopicolinate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

E1213-1G E1213-5G
TCE1213-1GEA 164.32 CAD
TCE1213-1G TCE1213-5G
Ethyl-6-aminopicolinate ≥98.0% (by GC, titration analysis)
Ethyl-6-aminopicolinate
Formula: C₈H₁₀N₂O₂
MDL Number: MFCD01009842
CAS Number: 69142-64-9

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 70.0 to 74.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR