Ethyl potassium oxalate ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

E1184-25G E1184-5G
TCE1184-25GEA 706.26 CAD
TCE1184-25G TCE1184-5G
Ethyl potassium oxalate ≥95.0% (by titrimetric analysis)
Ethyl potassium oxalate
Formula: C₄H₅KO₄
MDL Number: MFCD00045861
CAS Number: 1906-57-6
EINECS: 217-606-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR