3-Ethoxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-Ethoxy-4-fluorobenzeneboronic acid

E1183-1G
TCE1183-1GEA 132.72 CAD
TCE1183-1G
3-Ethoxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Ethoxy-4-fluorophenylboronic acid
Formula: C₈H₁₀BFO₃
MW: 183.97 g/mol
MDL Number: MFCD09475831
CAS Number: 900174-65-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR