trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (trans,trans)-4-(4-Ethoxy-2,3-difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane), trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-propylbicyclohexyl

E1157-1G E1157-5G
TCE1157-1GEA 87.53 CAD
TCE1157-1G TCE1157-5G
trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl
Formula: C₂₃H₃₄F₂O
MW: 364.521 g/mol
MDL Number: MFCD18433524
CAS Number: 123560-48-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C
Mesomorphic range 78.0 to 84.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR