Ethyl-2-formyl-3-oxopropionate ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Ethyl 2-formyl-3-oxopropanoate, Ethyl 2-Formyl-3-oxopropionate, Diformylacetic acid ethyl ester, 2-Formyl-3-oxopropionic acid ethyl ester, Ethyl diformylacetate

E1128-1G E1128-5G
TCE1128-1GEA 63.2 CAD
TCE1128-1G TCE1128-5G
Ethyl-2-formyl-3-oxopropionate ≥97.0% (by titrimetric analysis)
Ethyl-2-formyl-3-oxopropionate
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 32°C
Storage Temperature: Refrigerator
MDL Number: MFCD11112084
CAS Number: 80370-42-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR