Ethyl-4,6-dichloronicotinate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 4,6-dichloronicotinate

E1068-1G E1068-5G
TCE1068-1GEA 28.44 CAD
TCE1068-1G TCE1068-5G
Ethyl-4,6-dichloronicotinate ≥98.0% (by GC)
Ethyl-4,6-dichloronicotinate

CAS Number: 40296-46-6
MDL Number: MFCD00173839
Molecular Formula: C8H7Cl2NO2
Molecular Weight: 220.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 85
Melting point (°C): 33


Other Product Names / Synonyms: 4,6-Dichloronicotinic Acid Ethyl Ester, Ethyl 4,6-Dichloropyridine-3-carboxylate, 4,6-Dichloropyridine-3-carboxylic Acid Ethyl Ester

Formula: C₈H₇Cl₂NO₂
MW: 220.05 g/mol
Boiling Pt: 85 °C
Melting Pt: 33 °C
MDL Number: MFCD00234408
CAS Number: 40296-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc72b3e34336dd075426840f2d92d33c0adcc494&vid=fc72b3e34336dd075426840f2d92d33c0adcc494&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR