(-)-Epicatechin gallate ≥98.0% (by HPLC)

Supplier: TCI America

E0890-100MG E0890-20MG
TCE0890-100MGEA 1028.58 CAD
TCE0890-100MG TCE0890-20MG
(-)-Epicatechin gallate ≥98.0% (by HPLC)
(-)-Epicatechin gallate

CAS Number: 1257-08-5
MDL Number: MFCD00075936
Molecular Formula: C22H18O10
Molecular Weight: 442.38
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 255
Specific rotation [a]20/D: -177 deg (C=1, EtOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: (-)-cis-3,3',4',5,7-Pentahydroxyflavane 3-Gallate

Formula: C₂₂H₁₈O₁₀
Melting Pt: 255 °C
MDL Number: MFCD00075936
CAS Number: 1257-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0826ebda3f7bde29d199e9715a0882b95e868a3d&vid=0826ebda3f7bde29d199e9715a0882b95e868a3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR