2-Ethyl-3-hydroxy-6-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0833-5G
TCE0833-5GEA 96.38 CAD
TCE0833-5G
2-Ethyl-3-hydroxy-6-methylpyridine ≥98.0% (by GC, titration analysis)
2-Ethyl-3-hydroxy-6-methylpyridine

CAS Number: 2364-75-2
MDL Number: MFCD00462409
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 169


Product Categories: Bioscience, Antioxidants
Other Product Names / Synonyms: 2-Ethyl-6-methyl-3-pyridinol, Emoxypine

Formula: C₈H₁₁NO
Melting Pt: 169 °C
MDL Number: MFCD00462409
CAS Number: 2364-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e0f378b4d196fb294b665b5e74239e93fd86966&vid=0e0f378b4d196fb294b665b5e74239e93fd86966&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR