Ethyl-2,4,6-trimethylbenzoate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl Mesitylenecarboxylate, Ethyl 2,4,6-trimethylbenzoate, Mesitylenecarboxylic Acid Ethyl Ester, 2,4,6-Trimethylbenzoic Acid Ethyl Ester

E0736-25G
TCE0736-25GEA 605.14 CAD
TCE0736-25G
Ethyl-2,4,6-trimethylbenzoate ≥95.0%
Ethyl-2,4,6-trimethylbenzoate

CAS Number: 1754-55-8
MDL Number: MFCD00015439
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2,4,6-Trimethylbenzoic Acid Ethyl Ester, Ethyl Mesitylenecarboxylate, Mesitylenecarboxylic Acid Ethyl Ester

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 121 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00015439
CAS Number: 1754-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ed34d1b6e36277d64f716f9c2da03ec6bbc93bb&vid=5ed34d1b6e36277d64f716f9c2da03ec6bbc93bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR