Ethyl benzo[d]isoxazole-3-carboxylate ≥98.0%

Supplier: TCI America

E0731-100MG
TCE0731-100MGEA 71.1 CAD
TCE0731-100MG
Ethyl benzo[d]isoxazole-3-carboxylate ≥98.0%
Ethyl benzo[d]isoxazole-3-carboxylate

CAS Number: 57764-49-5
MDL Number: MFCD06659787
Molecular Formula: C10H9NO3
Molecular Weight: 191.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 59


Other Product Names / Synonyms: 1,2-Benzisoxazole-3-carboxylic Acid Ethyl Ester, 3-Ethoxycarbonyl-1,2-benzisoxazole

Formula: C₁₀H₉NO₃
Melting Pt: 59 °C
MDL Number: MFCD06659787
CAS Number: 57764-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23e3c9ffa8f0d4d80715a9fed88b4e7bcaad0a15&vid=23e3c9ffa8f0d4d80715a9fed88b4e7bcaad0a15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR