4-Ethylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

E0720-5G E0720-25G
TCE0720-5GEA 47.4 CAD
TCE0720-5G TCE0720-25G
4-Ethylphenylboronic acid (contains varying amounts of Anhydride)
4-Ethylphenylboronic acid

CAS Number: 63139-21-9
MDL Number: MFCD00859377
Molecular Formula: C8H11BO2
Molecular Weight: 149.98
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 141


Other Product Names / Synonyms: 4-Ethylbenzeneboronic Acid

Formula: C₈H₁₁BO₂
Melting Pt: 141 °C
MDL Number: MFCD00859377
CAS Number: 63139-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a90044ca8c78de716474d63d906cb1c3e3da9fd&vid=9a90044ca8c78de716474d63d906cb1c3e3da9fd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR