Ethyl-3-aminocrotonate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl 3-aminobut-2-enoate

E0709-25G
TCE0709-25GEA 143.78 CAD
TCE0709-25G
Ethyl-3-aminocrotonate ≥98.0% (by GC, titration analysis)
Ethyl 3-aminocrotonate

CAS Number: 7318-00-5
MDL Number: MFCD00008073
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 97
Freezing point (°C): 19
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 3-Amino-2-butenoic Acid Ethyl Ester

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Boiling Pt: 215 °C
Density: 1.02 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00008073
CAS Number: 7318-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a3aabde021eec7544de9d720228ef45aefb52a0&vid=1a3aabde021eec7544de9d720228ef45aefb52a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR