Ethyl stilbene-4-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Stilbene-4-carboxylic Acid Ethyl Ester, 4-(Ethoxycarbonyl)stilbene

E0678-1G E0678-5G
TCE0678-1GEA 133.83 CAD
TCE0678-1G TCE0678-5G
Ethyl stilbene-4-carboxylate ≥98.0%
Ethyl stilbene-4-carboxylate

CAS Number: 109463-48-1
MDL Number: MFCD06204340
Molecular Formula: C17H16O2
Molecular Weight: 252.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 110


Other Product Names / Synonyms: Stilbene-4-carboxylic Acid Ethyl Ester, 4-(Ethoxycarbonyl)stilbene

Formula: C₁₇H₁₆O₂
Melting Pt: 110 °C
MDL Number: MFCD06204340
CAS Number: 109463-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae652cc08792701179cb64f1c8e3c1603ea0773e&vid=ae652cc08792701179cb64f1c8e3c1603ea0773e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR