Ethyl-3-piperidinecarboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0584-5G E0584-25G
TCE0584-5GEA 34.76 CAD
TCE0584-5G TCE0584-25G
Ethyl-3-piperidinecarboxylate ≥98.0% (by GC, titration analysis)
Ethyl 3-piperidinecarboxylate

CAS Number: 5006-62-2
MDL Number: MFCD00005991
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 90
Specific Gravity (20/20): 1.02


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl Nipecotate, Nipecotic Acid Ethyl Ester, 3-Piperidinecarboxylic Acid Ethyl Ester

Formula: C₈H₁₅NO₂
Boiling Pt: 104 °C
Density: 1.02 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00005991
CAS Number: 5006-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=954dc618811ace6431a98b12f03b82d025046484&vid=954dc618811ace6431a98b12f03b82d025046484&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR