(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0452-5G E0452-1G
TCE0452-5GEA 909.13 CAD
TCE0452-5G TCE0452-001G
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide ≥98.0% (by GC, titration analysis)
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide

CAS Number: 114812-34-9
MDL Number: MFCD00191361
Molecular Formula: C7H14N2O
Molecular Weight: 142.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 110
Specific rotation [a]20/D: -117 deg (C=0.5, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acid Amides
Other Product Names / Synonyms: N-Ethyl-L-prolinamide

Formula: C₇H₁₄N₂O
Melting Pt: 110 °C
MDL Number: MFCD00191361
CAS Number: 114812-34-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edbbec08f637d356367f99c0855859ff83e0ae39&vid=edbbec08f637d356367f99c0855859ff83e0ae39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR