(Ethyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Ethyltriphenylphosphonium bromide

E0382-100G
TCE0382-100GEA 118.5 CAD
TCE0382-100G
(Ethyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
(Ethyl)triphenylphosphonium bromide

CAS Number: 1530-32-1
MDL Number: MFCD00011838
Molecular Formula: C20H20BrP
Molecular Weight: 371.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 210

Formula: C₂₀H₂₀BrP
MW: 371.26 g/mol
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00011838
CAS Number: 1530-32-1
EINECS: 216-223-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e5f23a90854325e4f985b04439980471e31fbac&vid=2e5f23a90854325e4f985b04439980471e31fbac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR