Sodium-8-ethoxyquinoline-5-sulfonate hydrate ≥98.0% (by HPLC)

Supplier: TCI America

E0338-5G E0338-25G
TCE0338-5GEA 105.86 CAD
TCE0338-5G TCE0338-025G
Sodium-8-ethoxyquinoline-5-sulfonate hydrate ≥98.0% (by HPLC)
Sodium-8-ethoxyquinoline-5-sulfonate hydrate

CAS Number: 7246-07-3
MDL Number: MFCD00006794
Molecular Formula: C11H11NO4S
Molecular Weight: 275.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: Actinoquinol Sodium Salt, 8-Ethoxyquinoline-5-sulfonic Acid Sodium Salt

Formula: C₁₁H₁₁NO₄S
MDL Number: MFCD00006794
CAS Number: 7246-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9cd5ae71e58d63036cd987c95f27d85a08c8964&vid=e532afade3fc1fe9d0ae805901e2001527f21113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR