2-(3,4-Epoxycyclohexyl)ethyltrimethoxysilane ≥97.0%

Supplier: TCI America

E0327-25ML E0327-500ML
TCE0327-025MLEA 49.82 CAD
TCE0327-025ML TCE0327-500ML
2-(3,4-Epoxycyclohexyl)ethyltrimethoxysilane ≥97.0%
2-(3,4-Epoxycyclohexyl)ethyltrimethoxysilane

CAS Number: 3388-04-3
MDL Number: MFCD00014485
Molecular Formula: C11H22O4Si
Molecular Weight: 246.38
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 310
Flash Point (°C): 141
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trimethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane

Formula: C₁₁H₂₂O₄Si
Boiling Pt: 310 °C
Density: 1.07 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00014485
CAS Number: 3388-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4614ddd56d6ca90a793e37608c151f02f3299e7c&vid=4614ddd56d6ca90a793e37608c151f02f3299e7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR