Ethylenediammonium dichloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Ethylenediamine dihydrochloride

E0078-500G E0078-25G
TCE0078-500GEA 216.46 CAD
TCE0078-500G TCE0078-025G
Ethylenediammonium dichloride ≥98.0% (by total nitrogen basis)
Ethylenediammonium dichloride

CAS Number: 333-18-6
MDL Number: MFCD00012524
Molecular Formula: C2H8N2
Molecular Weight: 133.02
Purity/Analysis Method: >98.0% (N)
Form: Crystal

Formula: C₂H₁₀Cl₂N₂
MW: 133.02 g/mol
MDL Number: MFCD00012524
CAS Number: 333-18-6
EINECS: 206-369-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49f7e5804a5a307745b33b255f86d65f9436556d&vid=49f7e5804a5a307745b33b255f86d65f9436556d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR