3,5-Lutidine-N-oxide ≥98.0% (by GC)

Supplier: TCI America
Warning

D5469-25G D5469-5G
TCD5469-25GEA 617.78 CAD
TCD5469-25G TCD5469-5G
3,5-Lutidine-N-oxide ≥98.0% (by GC)
3,5-Lutidine N-oxide
Formula: C₇H₉NO
Boiling Pt: 168°C
MDL Number: MFCD00126839
CAS Number: 3718-65-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR