Diclazuril ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(4-Chlorophenyl)-2-(2,6-dichloro-4-(3,5-dioxo-4,5-dihydro-1,2,4-triazin-2(3H)-yl)phenyl)acetonitrile, 2,6-Dichloro-alpha-(4-chlorophenyl)-4-[4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl]benzeneacetonitrile

D5457-1G D5457-5G
TCD5457-1GEA 85.32 CAD
TCD5457-1G TCD5457-5G
Diclazuril ≥98.0% (by HPLC, titration analysis)
Diclazuril
Formula: C₁₇H₉Cl₃N₄O₂
MW: 407.64 g/mol
MDL Number: MFCD00867203
CAS Number: 101831-37-2
Merck Index: 14,03080

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 295.0 to 299.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR