Dimethyl sulfoxide 99,0 GC_ASSAY_METHOD Spectrophotometry

Supplier: TCI America
Warning

Synonyms: Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane

D5293-100ML D5293-500ML
TCD5293-100MLEA 63.53 CAD
TCD5293-100ML TCD5293-500ML
Dimethyl sulfoxide 99,0 GC_ASSAY_METHOD Spectrophotometry
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Flash Pt: 95°C
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 14,03259

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Absorbance (270nm) max. 0.40
Absorbance (275nm) max. 0.25
Absorbance (280nm) max. 0.20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR