4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl ≥97.0% (by GC)

Supplier: TCI America

D5281-1G D5281-200MG
TCD5281-1GEA 714.16 CAD
TCD5281-1G TCD5281-200MG
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl ≥97.0% (by GC)
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl
Formula: C₂₄H₃₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD20257818
CAS Number: 2416-95-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 107.0 to 111.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR