9,9-Diphenyl-9H-9-silafluorene ≥98.0% (by GC)

Supplier: TCI America

D5034-1G D5034-200MG
TCD5034-1GEA 475.58 CAD
TCD5034-1G TCD5034-200MG
9,9-Diphenyl-9H-9-silafluorene ≥98.0% (by GC)
9,9-Diphenyl-9H-9-silafluorene
Formula: C₂₄H₁₈Si
CAS Number: 5550-08-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR