3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D5000-1G D5000-200MG
TCD5000-1GEA 240.16 CAD
TCD5000-1G TCD5000-200MG
3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(4-tert-butylphenyl)-9H-carbazole
Formula: C₂₂H₁₉Br₂N
MDL Number: MFCD12024283
CAS Number: 741293-42-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR