9,9-Di-n-octylfluorene-2,7-dicarboxaldehyde ≥98.0% (by GC)

Supplier: TCI America

D4671-1G
TCD4671-1GEA 538.78 CAD
TCD4671-1G
9,9-Di-n-octylfluorene-2,7-dicarboxaldehyde ≥98.0% (by GC)
9,9-Di-n-octylfluorene-2,7-dicarboxaldehyde
Formula: C₃₁H₄₂O₂
MW: 446.68 g/mol
CAS Number: 380600-91-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR