(4-Dimethylaminophenyl)di-tert-butylphosphine

Supplier: TCI America

D4531-1G D4531-5G
TCD4531-1GEA 137.46 CAD
TCD4531-1G TCD4531-5G
(4-Dimethylaminophenyl)di-tert-butylphosphine
(4-Dimethylaminophenyl)di-tert-butylphosphine

CAS Number: 932710-63-9
MDL Number: MFCD09265102
Molecular Formula: C16H28NP
Molecular Weight: 265.38
Form: Crystal
Color: White
Melting point (°C): 69


Other Product Names / Synonyms: 4-(Di-tert-butylphosphino)-N,N-dimethylaniline

Formula: C₁₆H₂₈NP
Melting Pt: 69 °C
MDL Number: MFCD09265102
CAS Number: 932710-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b3a1573f10701cbff91e11b0ea5f0ab546dadd9&vid=1b3a1573f10701cbff91e11b0ea5f0ab546dadd9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR