3,5-Dimethyl-1,2,4-triazole ≥95.0% (by GC, titration analysis)

Supplier: TCI America

D4481-1G D4481-5G
TCD4481-1GEA 79 CAD
TCD4481-1G TCD4481-5G
3,5-Dimethyl-1,2,4-triazole ≥95.0% (by GC, titration analysis)
3,5-Dimethyl-1,2,4-triazole

CAS Number: 7343-34-2
MDL Number: MFCD00656686
Molecular Formula: C4H7N3
Molecular Weight: 97.12
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 140

Formula: C₄H₇N₃
Boiling Pt: 260 °C
Melting Pt: 140 °C
MDL Number: MFCD00656686
CAS Number: 7343-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa885d8e9ae8633babcd9068a627e218012c3fc2&vid=aa885d8e9ae8633babcd9068a627e218012c3fc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR